Skip to Content
Merck
All Photos(3)

Documents

AV44234

Sigma-Aldrich

Anti-GUSB antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Glucuronidase, β, Anti-MPS7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

72 kDa

species reactivity

mouse, horse, guinea pig, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GUSB(2990)

General description

β-Glucuronidase (GUSB), a lysosomal enzyme, catalyzes the breakdown of complex carbohydrates by hydrolyzing β-D-glucuronic acid residues from the non-reducing end of mucopolysaccharides. GUSB is a stably expressed gene product that may be used to normalize the expression of other genes in processes such as mesenchymal stem cell differentiation.

Specificity

Anti-GUSB polyclonal antibody reacts with canine, bovine, zebrafish, chicken, human, mouse, rat, and pig β-glucuronidase proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human GUSB

Application

Anti-GUSB polyclonal antibody is used to tag β-glucuronidase protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe detect the presence of β-glucuronidase as a gene expression normalization reference protein.

Biochem/physiol Actions

GUSB plays an important role in the degradation of dermatan and keratan sulfates.

Sequence

Synthetic peptide located within the following region: VLGNKKGIFTRQRQPKSAAFLLRERYWKIANETRYPHSVAKSQCLENSPF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service