Skip to Content
Merck
All Photos(4)

Key Documents

AV43994

Sigma-Aldrich

Anti-SLC22A11 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC34282, Anti-OAT4, Anti-Solute carrier family 22 (organic anion/urate transporter), member 11, Anti-hOAT4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

60 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC22A11

Application

Anti-SLC22A11 (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

SLC22A11 (Organic Anion Transporter 4, OAT4) is a membrane protein involved in sodium-dependent transport and excretion of organic anions. This protein is expressed mainly in placenta and in the kidney. A variant of the gene encoding OAT4 has been implicated in the development of a major gout subtype called renal underexcretion type gout.

Sequence

Synthetic peptide located within the following region: ASSLVVLFFLPETQGLPLPDTIQDLESQKSTAAQGNRQEAVTVESTSL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James E Shima et al.
American journal of physiology. Renal physiology, 299(4), F767-F775 (2010-07-30)
Apical reabsorption from the urine has been shown to be important for such processes as the maintenance of critical metabolites in the blood and the excretion of nephrotoxic compounds. The solute carrier (SLC) transporter OAT4 (SLC22A11) is expressed on the
Masayuki Sakiyama et al.
Drug metabolism and pharmacokinetics, 29(2), 208-210 (2013-09-13)
Gout, a common disease, is a consequence of hyperuricemia, and increases the risks of hypertension, cardiovascular diseases, cerebrovascular diseases and renal failure. Gout can be classified into 3 types: the renal underexcretion (RUE) type, renal overload type and combined type.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service