Skip to Content
Merck
All Photos(3)

Documents

AV42487

Sigma-Aldrich

Anti-ASPN (AB3) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Aporin, Anti-FLJ20129, Anti-PLAP1, Anti-SLRR1C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASPN(54829)

General description

Asporin, periodontal ligament-associated protein 1 (PLAP1), is a member of the family of small leucine-rich proteoglycan (SLRP) family. Asporin is present in the cartilage extracellular matrix (ECM) where it plays a role in collagen mineralization. Asporin is involved in the mineralizaion of human dental pulp stem cells and predentin to dentin. Asporin inhibits bone morphogenetic protein-2 (BMP-2)-induced cytodifferentiation of periodontal ligament (PDL) cells and has been implicated in osteoarthritis.

The previously assigned protein identifier Q5TBF3 has been merged into Q9BXN1. Full details can be found on the UniProt database.

Specificity

Anti- PLAP1 (AB3) antibody reacts with canine, mouse, chicken, human, rat, and bovine Asporin (periodontal ligament-associated protein 1) proteins.

Immunogen

Synthetic peptide directed towards the middle region of human ASPN

Application

Anti- PLAP1 (AB3) antibody is used to tag asporin proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of asporin in cartilage mineralization and cytodifferentiation of cells such as periodontal ligament (PDL) cells.

Biochem/physiol Actions

ASPN belongs to a family of leucine-rich repeat (LRR) proteins associated with the cartilage matrix. The name asporin reflects the unique aspartate-rich N terminus and the overall similarity to decorin.ASPN belongs to a family of leucine-rich repeat (LRR) proteins associated with the cartilage matrix. The name asporin reflects the unique aspartate-rich N terminus and the overall similarity to decorin (MIM 125255) (Lorenzo et al., 2001).[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: NKISTVELEDFKRYKELQRLGLGNNKITDIENGSLANIPRVREIHLENNK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideki Kumagai et al.
Endocrine journal, 71(2), 139-152 (2024-01-04)
Nonalcoholic fatty liver disease (NAFLD) develops as a result of unhealthy lifestyle but improves with laparoscopic sleeve gastrectomy (LSG). The transforming growth factor (TGF)-β signaling pathway reportedly contributes to liver fibrosis, mainly in animal experiments. The aim of the present
Shengjie Wang et al.
Scientific reports, 7(1), 4112-4112 (2017-06-25)
Disc degeneration (DD) is a multifaceted chronic process that alters the structure and function of intervertebral discs. The pathophysiology of degeneration is not completely understood, but the consensus is that changes in genes encoding extracellular matrix (ECM) proteins in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service