Skip to Content
Merck
All Photos(2)

Key Documents

AV40475

Sigma-Aldrich

Anti-PRKRA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Protein kinase, interferon-inducible double stranded RNA-dependent activator

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

34 kDa

species reactivity

guinea pig, human, rat, rabbit, horse, mouse, dog, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKRA(8575)

General description

Protein kinase, interferon-inducible double stranded RNA-dependent activator/protein activator of the interferon-induced protein kinase (PRKRA, PACT) is a stress-modulated cellular activator of interferon (IFN)-induced double-stranded (ds) RNA-activated protein kinase (PKR) which is involved in antiviral defense in mammals. PACT and Mammalian Dicer interacts with double-stranded RNA-binding protein (TRBP) and associate with Dicer to stimulate cleavage of double-stranded RNA found in short hairpin and short interfering RNA (siRNA).

Specificity

Anti-PRKRA polyclonal antibody reacts with bovine, canine, human, mouse, and rat protein kinase, interferon-inducible double stranded RNA-dependent activators/protein activator of the interferon-induced protein kinases.

Immunogen

Synthetic peptide directed towards the middle region of human PRKRA

Application

Anti-PRKRA polyclonal antibody is used to tag protein kinase, interferon-inducible double stranded RNA-dependent activator/protein activator of the interferon-induced protein kinase for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of protein activator of the interferon-induced protein kinase (PACT) in stress response, antiviral activity and double-stranded RNA processing.

Biochem/physiol Actions

PRKRA contains 3 DRBM (double-stranded RNA-binding) domains. It appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor and activates EIF2AK2 in absence of double stranded RNA (dsRNA).

Sequence

Synthetic peptide located within the following region: RLPEYTLSQEGGPAHKREYTTICRLESFMETGKGASKKQAKRNAAEKFLA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service