Skip to Content
Merck
All Photos(3)

Key Documents

AV40417

Sigma-Aldrich

Anti-PSMA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HC2, Anti-MGC14542, Anti-MGC14575, Anti-MGC14751, Anti-MGC1667, Anti-MGC21459, Anti-MGC22853, Anti-Proteasome (prosome, macropain) subunit, α type, 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€386.00

€386.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€386.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€386.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

mouse, human, dog, rabbit, rat, horse, bovine, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PSMA1(5682)

General description

Proteasome (prosome, macropain) subunit, α type, 1 (PSMA1), a T1A family peptidase, is an α subunit of the complex 20S core structure of the proteosome, a multicatalytic proteinase complex.

Specificity

Anti-PSMA1 polyclonal antibody reacts with bovine, human, mouse, rat, chicken, zebrafish, and canine proteasome (prosome, macropain) subunit, α type, 1 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human PSMA1

Application

Anti-PSMA1 polyclonal antibody is used to tag pregnancy specific proteasome (prosome, macropain) subunit, α type, 1 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles proteasome (prosome, macropain) subunit, α type, 1 in proteasome structure and function.

Biochem/physiol Actions

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. PSMA1 is a member of the peptidase T1A family which is a 20S core alpha subunit.The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.

Sequence

Synthetic peptide located within the following region: TYLERHMSEFMECNLNELVKHGLRALRETLPAEQDLTTKNVSIGIVGKDL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jun He et al.
World journal of gastroenterology, 20(33), 11840-11849 (2014-09-11)
To investigate the molecular mechanisms of the anti-cancer activity of caffeic acid phenethyl ester (CAPE). Protein profiles of human colorectal cancer SW480 cells treated with or without CAPE were analysed using a two-dimensional (2D) electrophoresis gel-based proteomics approach. After electrophoresis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service