Skip to Content
Merck
All Photos(1)

Key Documents

AV36384

Sigma-Aldrich

Anti-DDX39 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DEAD (Asp-Glu-Ala-Asp) box polypeptide 39

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DDX39(10212)

Immunogen

Synthetic peptide directed towards the N terminal region of human DDX39

Biochem/physiol Actions

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX39 encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants encoding different isoforms.

Sequence

Synthetic peptide located within the following region: MAEQDVENDLLDYDEEEEPQAPQESTPAPPKKDIKGSYVSIHSSGFRDFL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Kubota et al.
Japanese journal of clinical oncology, 42(8), 730-741 (2012-06-23)
This study aimed to validate two prognostic biomarkers, pfetin and adenosine triphosphate-dependent RNA helicase DDX39 (DDX39), in gastrointestinal stromal tumour. Prognostic biomarkers have long been required for the optimal use of kinase inhibitors in gastrointestinal stromal tumour. The expression level
Takeyuki Sugiura et al.
Cancer biology & therapy, 6(6), 957-964 (2007-06-06)
To explore differentially expressed genes involved in non-small cell lung cancer progression, we used the gene expression profile database of various human tissues and identified DDX39, a new member of the DEAD box RNA helicases, showing overexpression in human lung
Frances V Fuller-Pace
Nucleic acids research, 34(15), 4206-4215 (2006-08-29)
The DExD/H box family of proteins includes a large number of proteins that play important roles in RNA metabolism. Members of this family have been shown to act as RNA helicases or unwindases, using the energy from ATP hydrolysis to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service