Skip to Content
Merck
All Photos(3)

Key Documents

AV36244

Sigma-Aldrich

Anti-ZFP42 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Zinc finger protein 42 homolog (mouse)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFP42(132625)

Immunogen

Synthetic peptide directed towards the N terminal region of human ZFP42

Biochem/physiol Actions

ZFP42, also known as REX1 and ZNF754, is a zinc finger protein and a marker of pluripotent stem cells. It is expressed in placenta during spermatogenesis and early embryogenesis. ZFP42 regulates the expression of endogenous retroviral elements in mouse embryonic stem cells.[1]

Sequence

Synthetic peptide located within the following region: MSQQLKKRAKTRHQKGLGGRAPSGAKPRQGKSSQDLQAEIEPVSAVWALC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Guallar et al.
Nucleic acids research, 40(18), 8993-9007 (2012-07-31)
Rex1/Zfp42 is a Yy1-related zinc-finger protein whose expression is frequently used to identify pluripotent stem cells. We show that depletion of Rex1 levels notably affected self-renewal of mouse embryonic stem (ES) cells in clonal assays, in the absence of evident
Naira C Rezende et al.
Developmental biology, 356(2), 370-382 (2011-06-07)
Rex1 (Zfp42), GeneID 132625, is a gene whose expression is closely associated with pluripotency/multipotency in both mouse and human embryonic stem cells. To study the function of the murine Rex1 gene in vivo, we have used cre/lox technology to create

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service