Skip to Content
Merck
All Photos(6)

Key Documents

AV34673

Sigma-Aldrich

Anti-CREB3L2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-cAMP responsive element binding protein 3-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€289.00

€289.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€289.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€289.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREB3L2(64764)

General description

RCOR2 is a part of the LSD1 complex and is known to modulate ESC properties. RCOR2 also substitutes for SOX2 during somatic cell reprogramming.
Rabbit Anti-RCOR2 antibody recognizes canine, bovine, human, mouse, and rat RCOR2.

Immunogen

Synthetic peptide directed towards the N terminal region of human CREB3L2

Application

Rabbit Anti-RCOR2 antibody is suitable for western blot applications at 1.25 μg/ml and for IHC of praffin-embedded tissue sections at 4-8 μg/ml.

Biochem/physiol Actions

CREB3L2 is a member of the old astrocyte specifically induced substance (OASIS) DNA binding and basic leucine zipper dimerization (bZIP) family of transcription factors, which includes CREB3 (MIM 606443) and CREB4 (MIM 607138).

Sequence

Synthetic peptide located within the following region: HSYSLCEEPRAQSPFTHITSDSFNDDEVESEKWYLSTDF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Yang et al.
Stem cells (Dayton, Ohio), 29(5), 791-801 (2011-03-25)
Histone demethylase LSD1 can form complex with different Rcor family corepressors in different cell types. It remains unknown if cell-specific Rcor proteins function specifically in distinct cell types. Here, we report that Rcor2 is predominantly expressed in ESCs and forms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service