Skip to Content
Merck
All Photos(3)

Key Documents

AV34376

Sigma-Aldrich

Anti-ENO1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Enolase 1, (α)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€308.00

€308.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€308.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€308.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

guinea pig, rat, rabbit, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ENO1(2023)

General description

α-enolase (Eno1) is one of three enolases. Each is composed of 2 α, 2 gamma, or 2 β subunits, and functions as a glycolytic enzyme. Eno-1 is also known as MPB-1.

The previously assigned protein identifier Q53HR3 has been merged into P06733. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human ENO1

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ENO1 is one of three enolase isoenzymes found in mammals; it encodes alpha-enolase, a homodimeric soluble enzyme, and also encodes a shorter monomeric structural lens protein, tau-crystallin. The two proteins are made from the same message. The full length protein, the isoenzyme, is found in the cytoplasm. The shorter protein is produced from an alternative translation start, is localized to the nucleus, and has been found to bind to an element in the c-myc promoter. A pseudogene has been identified that is located on the other arm of the same chromosome.

Sequence

Synthetic peptide located within the following region: VVGLCTGQIKTGAPCRSERLAKYNQLLRIEEELGSKAKFAGRNFRNPLAK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Misako Sato et al.
PloS one, 10(5), e0126483-e0126483 (2015-05-21)
Transforming growth factor-β (TGF-β) has a dual role in tumorigenesis, acting as either a tumor suppressor or as a pro-oncogenic factor in a context-dependent manner. Although TGF-β antagonists have been proposed as anti-metastatic therapies for patients with advanced stage cancer
Michael Lutter et al.
Biological psychiatry, 81(9), 770-777 (2016-11-26)
While eating disorders (EDs) are thought to result from a combination of environmental and psychological stressors superimposed on genetic vulnerability, the neurobiological basis of EDs remains incompletely understood. We recently reported that a rare missense mutation in the gene for
Hang Yin et al.
Gastroenterology research and practice, 2018, 3842198-3842198 (2018-02-28)
We investigated in this study the expression of ENO1 in tissues and plasma of PDAC patients to evaluate its clinicopathological and diagnostic significance. ENO1 protein expression was detected in tissue microarray of human PDAC and adjacent noncancer tissues. Electrochemiluminescence immunoassay

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service