Skip to Content
Merck
All Photos(4)

Key Documents

AV32697

Sigma-Aldrich

Anti-MEOX1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Mesenchyme homeobox 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEOX1(4222)

General description

MEOX1 is a homeobox transcription factor that regulates somite development. It is known to maintain sclerotome polarity and is also involved in axial skeleton formation. MEOX1 mutations have been linked to Klippel-Feil syndrome (KFS).
Rabbit Anti-MEOX1 bovine, canine, and human MEOX1.

Immunogen

Synthetic peptide directed towards the N terminal region of human MEOX1

Application

Rabbit Anti-MEOX1 can be used for western blot applications at a concentration of 0.2-2.0μg/ml. It can also be used for IHC at 4-8μg/ml using paraffin-embedded tissues.

Biochem/physiol Actions

MEOX1 belongs to a family of nonclustered, diverged homeobox genes. It may play a role in regulating growth and differentiation.

Sequence

Synthetic peptide located within the following region: YPPTPFSFHQKPDFLATATAAYPDFSASCLAATPHSLPQEEHIFTEQHPA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutation in MEOX1 gene causes a recessive Klippel-Feil syndrome subtype.
Bayrakli F, et al. et al.
BMC Genetics (2013)
Susan Skuntz et al.
Developmental biology, 332(2), 383-395 (2009-06-13)
Meox1 and Meox2 are two related homeodomain transcription factor genes that together are essential for the development of all somite compartments. Here we show that mice homozygous for Meox1 mutations alone have abnormalities that are restricted to the sclerotome and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service