Synthetic peptide directed towards the N terminal region of human KLF15
Application
Rabbit Anti-KLF15 antibody can be used for western blot applications at a concentration of 1.0-2.0μg/ml. It can also be used for immunohistochemical applications at a concentration of 4-8μg/ml using paraffin-embedded tissues.
Biochem/physiol Actions
KLF15 is a Cys2-His2 zinc finger gene. It is found abundantly expressed in the liver, kidneys, heart, and skeletal muscle.
Sequence
Synthetic peptide located within the following region: DASSPCSCSSPDSQALCSCYGGGLGTESQDSILDFLLSQATLGSGGGSGS
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Hypertrophic cardiomyopathy (HCM) is the most common genetic disease of the cardiac muscle, frequently caused by mutations in MYBPC3. However, little is known about the upstream pathways and key regulators causing the disease. Therefore, we employed a multi-omics approach to
Molecular medicine reports, 21(3), 1336-1345 (2020-01-11)
The aim of the present study was to investigate the protective effect of Kruppel‑like factor 15 (KLF15) overexpression on heart failure (HF) induced by left ventricular (LV) pressure overload in mice. Wild‑type (WT) mice and cardiac‑specific KLF15‑overexpressed transgenic (TG) mice
Journal of cellular and molecular medicine, 25(12), 5691-5706 (2021-05-06)
Mesangial cell (MC) proliferation is a key pathological feature in a number of common human renal diseases, including mesangial proliferative nephritis and diabetic nephropathies. Knowledge of MC responses to pathological stimuli is crucial to the understanding of these disease processes.
International journal of molecular medicine, 42(3), 1593-1602 (2018-06-15)
Krüppel‑like factor 15 (KLF15), also known as kidney‑enriched transcription factor, is known to participate in podocyte differentiation. However, the role of KLF15 in chronic podocyte injury remains incompletely understood, particularly in proteinuric disease models. In the present study, the 5/6 nephrectomy
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.