Skip to Content
Merck
All Photos(1)

Key Documents

AV32066

Sigma-Aldrich

Anti-IRX4 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Iroquois homeobox protein 4, Anti-MGC131996

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

pig, bovine, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IRX4(50805)

General description

IRX4 gene variations have been implicated in congenital heart disease and prostate cancer susceptibility. Rabbit Anti-IRX4 antibody recognizes bovine, human, mouse, and rat IRX4.

Immunogen

Synthetic peptide directed towards the N terminal region of human IRX4

Application

Rabbit Anti-IRX4 antibody can be used for western blot applications at a concentration of 2.5μg/ml.

Biochem/physiol Actions

IRX4 is likely to be an important mediator of ventricular differentiation during cardiac development.

Sequence

Synthetic peptide located within the following region: SAAALGVYGGPYGGSQGYGNYVTYGSEASAFYSLNSFDSKDGSGSAHGGL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi Cheng et al.
Human genetics, 130(5), 657-662 (2011-05-06)
IRX4 was the first identified cardiac transcription factor that is restricted to the ventricles at all stages of heart development. Irx4-deficient mice show ventricular dysfunction and develop cardiomyopathy. To study the potential impact of sequence variations in IRX4 on congenital
Hai Ha Nguyen et al.
Human molecular genetics, 21(9), 2076-2085 (2012-02-11)
Recent genome-wide association studies (GWAS) identified a number of prostate cancer (PC) susceptibility loci, but most of their functional significances are not elucidated. Through our previous GWAS for PC in a Japanese population and subsequent resequencing and fine mapping, we
Takao Kitagawa et al.
PloS one, 17(10), e0269077-e0269077 (2022-10-05)
Ewing's sarcoma is the second most common bone malignancy in children or young adults and is caused by an oncogenic transcription factor by a chromosomal translocation between the EWSR1 gene and the ETS transcription factor family. However, the transcriptional mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service