Skip to Content
Merck
All Photos(2)

Key Documents

AV31259

Sigma-Aldrich

Anti-CEBPB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CCAAT/enhancer binding protein (C/EBP), β

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

36 kDa

species reactivity

dog, guinea pig, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CEBPB(1051)

General description

CEBPB is a hepatic transcription factor that contains the basic leucine zipper (bZIP) domain. This transcription factor is known to interact with genes involved in acute phase response, metabolic activities, homeostatic functions, and the progression of cell cycle[1].
Rabbit Anti-CEBPB antibody binds to bovine, human, and pig CEBPB.

Immunogen

Synthetic peptide directed towards the C terminal region of human CEBPB

Application

Rabbit Anti-CEBPB antibody can be used for western blot applications at 0.5μg/ml.

Biochem/physiol Actions

The protein encoded by this intronless gene, CEBPB, is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related proteins CEBP-alpha, CEBP-delta, and CEBP-gamma. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses and has been shown to bind to the IL-1 response element in the IL-6 gene, as well as to regulatory regions of several acute-phase and cytokine genes. In addition, the encoded protein can bind the promoter and upstream element and stimulate the expression of the collagen type I gene.

Sequence

Synthetic peptide located within the following region: ADAKAPPTACYAGAAPAPSQVKSKAKKTVDKHSDEYKIRRERNNIAVRKS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janus Schou Jakobsen et al.
Genome research, 23(4), 592-603 (2013-02-14)
Dynamic shifts in transcription factor binding are central to the regulation of biological processes by allowing rapid changes in gene transcription. However, very few genome-wide studies have examined how transcription factor occupancy is coordinated temporally in vivo in higher animals.
Fen Wang et al.
Hypertension (Dallas, Tex. : 1979), 63(2), 281-288 (2013-11-06)
DNA methylation patterns are maintained in adult somatic cells. Recent findings, however, suggest that all methylation patterns are not preserved. We demonstrate that stimulatory signals can change the DNA methylation status at a CCAAT/enhancer binding protein (CEBP) binding site and
Philip D Campbell et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(44), 14717-14732 (2014-10-31)
Mutations in Kinesin proteins (Kifs) are linked to various neurological diseases, but the specific and redundant functions of the vertebrate Kifs are incompletely understood. For example, Kif5A, but not other Kinesin-1 heavy-chain family members, is implicated in Charcot-Marie-Tooth disease (CMT)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service