Skip to Content
Merck
All Photos(2)

Key Documents

AV31085

Sigma-Aldrich

Anti-ELF1 (AB2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-E74-like factor 1 (ets domain transcription factor)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

67 kDa

species reactivity

rat, bovine, dog, human, guinea pig, rabbit, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELF1(1997)

General description

ELF1 is known to function as a transcription factor that regulates the Tie2 gene during blood vessel development. Elf1 has also been implicated in the regulation of terminal transferase gene.
Rabbit Anti-ELF1 (AB2) antibody recognizes rat, human, bovine, canine, and mouse ELF1.

Immunogen

Synthetic peptide directed towards the N terminal region of human ELF1

Application

Rabbit Anti-ELF1 (AB2) antibody is suitable for use in western blot (1μg/ml) applications.

Biochem/physiol Actions

ELF1 belongs to the ETS family. It contains 1 ETS DNA-binding domain. ELF1 is a transcription factor that activates the LYN and BLK promoters. It appears to be required for the T-cell-receptor-mediated trans activation of HIV-2 gene expression. ELF1 binds specifically to two purine-rich motifs in the HIV-2 enhancer.

Sequence

Synthetic peptide located within the following region: VTLDGIPEVMETQQVQEKYADSPGASSPEQPKRKKGRKTKPPRPDSPATT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Ernst et al.
Molecular and cellular biology, 16(11), 6121-6131 (1996-11-01)
The terminal deoxynucleotidyltransferase (TdT) gene represents an attractive model for the analysis of gene regulation during an early phase of lymphocyte development. In previous studies, we identified a DNA element, termed D', which is essential for TdT promoter activity in
A Dube et al.
Circulation research, 88(2), 237-244 (2001-02-07)
Vascular development requires the tightly coordinated expression of several growth factors and their receptors. Among these are the Tie1 and Tie2 receptors, which are almost exclusively endothelial cell-specific. The critical transcriptional regulators of vascular-specific gene expression remain largely unknown. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service