Skip to Content
Merck
All Photos(4)

Key Documents

AV30037

Sigma-Aldrich

Anti-GLIS2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-GLIS family zinc finger 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

dog, human, horse, bovine, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLIS2(84662)

General description

GLIS family zinc finger 2 (GLIS2), a Krüppel-like zinc finger protein family member with transactivation and repressor functions, is involved in kidney development, the maintenance of normal renal function and neurogenesis. Glis2 interacts with β-catenin and may function as a negative modulator of β-catenin/TCF-mediated transcription. Defective GLIS2 has been linked to the development of nephronophthisis.
Rabbit polyclonal anti-GLIS2 antibody reacts with canine, human, chicken, rat, bovine, and mouse GLIS family zinc finger 2 transcription factors.

Immunogen

Synthetic peptide directed towards the N terminal region of human GLIS2

Application

Rabbit polyclonal anti-GLIS2 antibody is used to tag GLIS family zinc finger 2 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of GLIS family zinc finger 2 in kidney development and maintenance. Anti-GLIS2 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

Members of the Kruppel-like zinc finger protein family, such as GLIS2, function as activators and/or repressors of gene transcription.

Sequence

Synthetic peptide located within the following region: QDLVDHVNDYHVKPEKDAGYCCHWEGCARHGRGFNARYKMLIHIRTHTNE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service