Skip to Content
Merck
All Photos(1)

Key Documents

AV13101

Sigma-Aldrich

Anti-GABRG3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-γ-Aminobutyric acid (GABA) A receptor, γ 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GABRG3(2567)

Immunogen

Synthetic peptide directed towards the N terminal region of human GABRG3

Application

Anti-GABRG3 (AB2) antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

GABRG3 is a subunit of GABAA receptor, the main inhibitory receptor in the brain.[1] Polymorphisms of GABRG3 gene result in altered responsiveness to GABA and may be involved in alcohol dependence.[2]

Sequence

Synthetic peptide located within the following region: LHARSRKVEEDEYEDSSSNQKWVLAPKSQDTDVTLILNKLLREYDKKLRP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle M Dick et al.
Alcoholism, clinical and experimental research, 28(1), 4-9 (2004-01-28)
Evidence from human, animal, and in vitro cell models suggests that gamma-aminobutyric acid (GABA), the major inhibitory neurotransmitter in the human central nervous system, is involved in many of the neurochemical pathways that affect alcohol use, abuse, and dependence. Both
H Hörtnagl et al.
Neuroscience, 236, 345-372 (2013-01-23)
The GABAA receptor is the main inhibitory receptor in the brain and its subunits originate from different genes or gene families (α1-α6, β1-β3, γ1-γ3, δ, ε, θ, π, or ρ1-3). In the mouse brain the anatomical distribution of GABAA receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service