Skip to Content
Merck
All Photos(1)

Key Documents

AV13019

Sigma-Aldrich

Anti-CHRNA9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cholinergic receptor, nicotinic, α-9 (muscle)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€374.00

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€374.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

guinea pig, horse, human, bovine, dog, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CHRNA9(55584)

Immunogen

Synthetic peptide directed towards the N terminal region of human CHRNA9

Application

Anti-CHRNA9 antibody produced in rabbit is suitable for western blotting at a concentration of 0.0625 μg/ml.

Biochem/physiol Actions

CHRNA9 is a transmembrane oligomeric ligand-gated nicotinic receptor that induces ion channel opening for the movement of positive ions when it is activated by cholinergic binding. Nicotinic acetylcholine receptors mediate presynaptic, postsynaptic and extrasynaptic signaling.[1] The expression of CHRNA9 receptor has been observed to be elevated in human breast epithelial cells during tumorigenesis.[2] CHRNA9 has been implicated in stress-induced functional plasticity of rat adrenal medulla.[3]

Sequence

Synthetic peptide located within the following region: MNWSHSCISFCWIYFAASRLRAAETADGKYAQKLFNDLFEDYSNALRPVE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Hwa Lee et al.
Journal of the National Cancer Institute, 102(17), 1322-1335 (2010-08-25)
Large epidemiological cohort studies in the United States have indicated that active and passive smoking are associated with increased breast cancer risk. However, there was no direct evidence of an effect of tobacco carcinogens on the cellular molecules involved in
Claude Colomer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 30(19), 6732-6742 (2010-05-14)
An increase in circulating adrenal catecholamine levels constitutes one of the mechanisms whereby organisms cope with stress. Accordingly, stimulus-secretion coupling within the stressed adrenal medullary tissue undergoes persistent remodeling. In particular, cholinergic synaptic neurotransmission between splanchnic nerve terminals and chromaffin
Inmaculada Posadas et al.
Current neuropharmacology, 11(3), 298-314 (2013-11-02)
Many studies have focused on expanding our knowledge of the structure and diversity of peripheral and central nicotinic receptors. Nicotinic acetylcholine receptors (nAChRs) are members of the Cys-loop superfamily of pentameric ligand-gated ion channels, which include GABA (A and C)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service