Skip to Content
Merck
All Photos(2)

Key Documents

AV100811

Sigma-Aldrich

Anti-GTF2H1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-General transcription factor IIH, polypeptide 1, 62 kDa

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€327.00

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

guinea pig, human, rat, mouse, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GTF2H1(2965)

Immunogen

Synthetic peptide directed towards the N terminal region of human GTF2H1

Application

Anti-GTF2H1 antibody is suitable for western blotting at a concentration of 1-2 μg/ml. For immunohistochemistry of paraffin-embedded tissue sections, a concentration of 4-8 μg/ml is suitable.

Biochem/physiol Actions

GTF2H1 is RNA polymerase II basal transcription factor that is involved in DNA repair, transcription elongation and cell cycle.[1]

Sequence

Synthetic peptide located within the following region: EEVLLIVKKVRQKKQDGALYLMAERIAWAPEGKDRFTISHMYADIKCQKI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximiliano M Portal
Proceedings of the National Academy of Sciences of the United States of America, 108(21), 8686-8691 (2011-05-12)
General transcription factor IIH (TFIIH) is a complex RNA polymerase II basal transcription factor comprising 10 different polypeptides that display activities involved in transcription and DNA repair processes. Although biochemical studies have uncovered TFIIH importance, little is known about how
D Patschan et al.
American journal of physiology. Renal physiology, 307(6), F686-F694 (2014-08-01)
Diabetic nephropathy is the most frequent single cause of end-stage renal disease in our society. Microvascular damage is a key event in diabetes-associated organ malfunction. Early endothelial outgrowth cells (eEOCs) act protective in murine acute kidney injury. The aim of
Jumpei Kashima et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 44(7), 779-787 (2013-06-19)
Autophagy has been implicated in lipid droplet (LD) turnover. Adipose differentiation-related protein (ADRP) and microtubule-associated protein 1 light chain 3 (LC3) monitor LD and autophagosomes, respectively. We examined whether immunohistochemical staining of ADRP and LC3 can monitor LD and autophagy
Li Wang et al.
Biochemical and biophysical research communications, 448(4), 448-453 (2014-05-07)
Interleukin-10 is an important cytokine that regulates immune response. Previous studies have shown that human cytomegalovirus can trigger cell autophagy during the early stages of infection. To our knowledge, whether IL-10 inhibits HCMV-induced autophagy and virus replication has not been
A Trocoli et al.
Cell death and differentiation, 21(12), 1852-1861 (2014-07-19)
The p62/SQSTM1 adapter protein has an important role in the regulation of several key signaling pathways and helps transport ubiquitinated proteins to the autophagosomes and proteasome for degradation. Here, we investigate the regulation and roles of p62/SQSTM1 during acute myeloid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service