Skip to Content
Merck
All Photos(1)

Documents

AQ0144

Sigma-Aldrich

AQUA Peptide VYIHPFHLVIHDESTCEQLA[K]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10713C6H171N2715N2O32S
Molecular Weight:
2487.75
UNSPSC Code:
12352200

form

solid

mol wt

monoisotopic mol wt 2486.25 Da

shipped in

wet ice

storage temp.

−20°C

Application

Protein-AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School. Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.
This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company.  The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration.  The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party.  Information about licenses for excluded uses is available from:  E. I. du Pont de Nemours and Company;  Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
AQUA is a trademark of Harvard Medical School
Protein AQUA is a trademark of Harvard Medical School

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service