Skip to Content
Merck

Skip To

APPA014

HIV Protease Activity Detection Kit

Synonym(s):

Cut-N-Glow HIV Protease Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Check Cart for Availability
€ 972,00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200

€ 972,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shipped in

dry ice

storage temp.

−20°C

Application

This kit is specific for the HIV Protease, a retroviral aspartyl protease that is critical to viral life-cycle. The Cut-N-Glow approach involves the introduction of a structural distortion into one of the complementary fragments (GFP 11), through the use of a conditionally stable tether, which serves to constrain the N and C termini of GFP 11 closely in space, thereby, diminishing the mutual affinity of the two fragments and blocking protein self-assembly until the tether is cleaved. The distortion can be reversed upon site-specific proteolysis of the tether, resulting in GFP 11 fragment assembly with GFP 1-10, generating reconstituted, functional GFP. Depending on the tether, the chimeric GFP can serve as a substrate for proteases from the three major classes: serine, cysteine, and aspartic acid.
This kit contains sufficient reagents for one 96 well plate (96 tests)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CS0010ET0300ET0200
storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

form

solution

form

-

form

-

form

-

shipped in

dry ice

shipped in

wet ice

shipped in

-

shipped in

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200


flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters