AC-0350
Neuron-specific enolase (NSE) (EP319) Rabbit Monoclonal Primary Antibody
About This Item
biological source
rabbit
Quality Level
antibody form
Ig fraction of antiserum
clone
monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic
General description
Quality
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Physical form
Other Notes
For Technical Service please contact: 800-665-7284 or email: [email protected]
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service