Skip to Content
Merck

Skip To

A9971

2-Aminobenzoyl-Ala-Phe-Ala-Phe-Asp-Val-Phe-Tyr(NO2)-Asp

powder, >90% (HPLC)

Synonym(s):

Anthraniloyl-Ala-Phe-Ala-Phe-Asp-Val-Phe-3-nitro-Tyr-Asp-OH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 451,00

About This Item

Empirical Formula (Hill Notation):
C62H71N11O18
CAS Number:
Molecular Weight:
1258.29
UNSPSC Code:
12352204
NACRES:
NA.32
MDL number:

€ 451,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2-Aminobenzoyl-Ala-Phe-Ala-Phe-Asp-Val-Phe-Tyr(NO2)-Asp, >90% (HPLC)

Quality Segment

assay

>90% (HPLC)

form

powder

mol wt

1258.31

solubility

1% ammonia: soluble 1 mg/mL, clear, yellow

storage temp.

−20°C

Application

Fluorogenic substrate for Asp-specific proteases from Staphylococcus aureus, Bacillus lichenformis, and Streptomyces griseus.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCP0262A3401B4310
assay

>90% (HPLC)

assay

≥99% (TLC)

assay

≥98% (TLC)

assay

≥95% (HPLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

solid

solubility

1% ammonia: soluble 1 mg/mL, clear, yellow

solubility

methanol: ~50 mg/mL, almost clear, almost colorless

solubility

ethanol: 20 mg/mL, clear, colorless to light yellow

solubility

H2O: >1 mg/mL

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

mol wt

1258.31

mol wt

370.41

mol wt

-

mol wt

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters