Skip to Content
Merck

Skip To

A9834

2-Amino-5,6-dihydro-6-methyl-4H-1,3-thiazine

≥98%

Synonym(s):

AMT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
€ 358,00

About This Item

Empirical Formula (Hill Notation):
C5H10N2S · HCl
CAS Number:
Molecular Weight:
166.67
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:

€ 358,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥98%

form

powder

solubility

water: 25 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

Cl[H].CC1CCN=C(N)S1

InChI

1S/C5H10N2S.ClH/c1-4-2-3-7-5(6)8-4;/h4H,2-3H2,1H3,(H2,6,7);1H

InChI key

HVJCRMIQAMEJNM-UHFFFAOYSA-N

Gene Information

human ... FPGS(2356)

Biochem/physiol Actions

As a selective type II (inducible) nitric oxide synthase (NOS) inhibitor, AMT was tested against LPS-induced inflammation in rats.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M6500C13408H7654
biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

synthetic (organic)

assay

≥98%

assay

≥98% (titration)

assay

98%

assay

≥98% (TLC)

Gene Information

human ... FPGS(2356)

Gene Information

-

Gene Information

-

Gene Information

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

-

form

powder

form

powder or crystals

form

crystalline powder, powder or crystals

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters