Skip to Content
Merck
All Photos(1)

Key Documents

A9755

Sigma-Aldrich

Androsterone

≥99%

Synonym(s):

3α-Hydroxy-5α-androstan-17-one, 5α-Androstan-3α-ol-17-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H30O2
CAS Number:
Molecular Weight:
290.44
Beilstein:
2217626
MDL number:
UNSPSC Code:
51111800

Assay

≥99%

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

mp

181-184 °C (lit.)

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CCC(=O)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@@H](O)C2

InChI

1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12-16,20H,3-11H2,1-2H3/t12-,13+,14-,15-,16-,18-,19-/m0/s1

InChI key

QGXBDMJGAMFCBF-HLUDHZFRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anabolic steroid

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Su et al.
Endocrinology, 140(11), 5275-5284 (1999-10-28)
17Beta-hydroxysteroid dehydrogenases (17betaHSDs) convert androgens and estrogens between their active and inactive forms, whereas retinol dehydrogenases catalyze the conversion between retinol and retinal. Retinol dehydrogenases function in the visual cycle, in the generation of the hormone retinoic acid, and some
X Chai et al.
The Journal of biological chemistry, 272(52), 33125-33131 (1998-01-31)
We report a mouse cDNA that encodes a 317-amino acid short-chain dehydrogenase which recognizes as substrates 9-cis-retinol, 11-cis-retinol, 5alpha-androstan-3alpha,17beta-diol, and 5alpha-androstan-3alpha-ol-17-one. This cis-retinol/androgen dehydrogenase (CRAD) shares closest amino acid similarity with mouse retinol dehydrogenase isozymes types 1 and 2 (86
S H Doss et al.
Archives of pharmacal research, 22(5), 496-501 (1999-11-05)
The reaction of epi-androsterone with cyanoacetyl hydrazine (2) gave the hydrazone derivative 3. The reactivity of 3 towards chemical reagents was studied to give pyridine, thiazole and thiophene derivatives.
Guy-Bertrand Djigoue et al.
Acta crystallographica. Section C, Crystal structure communications, 68(Pt 6), o231-o234 (2012-06-07)
The title compounds, (3R,5S,5'R,8R,9S,10S,13S,14S)-10,13-dimethyl-5'-(2-methylpropyl)tetradecahydro-6'H-spiro[cyclopenta[a]phenanthrene-3,2'-[1,4]oxazinane]-6',17(2H)-dione, C(26)H(41)NO(3), (I), and methyl (2R)-2-[(3R,5S,8R,9S,10S,13S,14S)-10,13-dimethyl-2',17-dioxohexadecahydro-3'H-spiro[cyclopenta[a]phenanthrene-3,5'-[1,3]oxazolidin-3'-yl]]-4-methylpentanoate, C(28)H(43)NO(5), (II), possess the typical steroid shape (A-D rings), but they differ in their extra E ring. The azalactone E ring in (I) shows a half-chair conformation, while the carbamate E
N Duijvesteijn et al.
Journal of animal science, 90(8), 2465-2475 (2012-03-01)
In the pig industry, male piglets are surgically castrated early in life to prevent boar taint. Boar taint is mainly caused by androstenone and skatole. Androstenone is a pheromone that can be released from the salivary glands when the boar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service