Skip to Content
Merck
All Photos(1)

Key Documents

A9630

Sigma-Aldrich

4-Androstene-3,17-dione

≥98%

Synonym(s):

Androstenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
Beilstein:
2059239
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥98%

drug control

USDEA Schedule III; Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

mp

170-171 °C (lit.)

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)C(=O)CC[C@@]24[H]

InChI

1S/C19H26O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-16H,3-10H2,1-2H3/t14-,15-,16-,18-,19-/m0/s1

InChI key

AEMFNILZOJDQLW-QAGGRKNESA-N

Gene Information

human ... HSD17B3(3293)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Testosterone precursor and metabolite with androgenic activity.
Testosterone precursor and metabolite with androgenic activity; may increase the risk of prostate or pancreatic cancer.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R S Riikonen
Journal of child neurology, 11(5), 383-388 (1996-09-01)
Infants with cryptogenic infantile spasms seem to differ from those with symptomatic spasms in having a higher cerebrospinal fluid corticotropin content, different levels of corticotropin release after exogenous vasopressin, higher serum levels of progesterone, higher dehydroepiandrosterone: androstenedione ratio (during corticotropin
A Mouritsen et al.
The Journal of clinical endocrinology and metabolism, 98(9), 3755-3764 (2013-07-05)
Little is known about the possible deleterious effects of phthalate exposure on endogenous sex steroid levels in children. Our objective was to investigate whether urinary phthalate metabolite levels are associated with circulating adrenal androgen levels and age at puberty. This
P S Feldman et al.
Cancer, 49(4), 714-721 (1982-02-15)
A 51-year-old man had a malignant Leydig cell tumor of the testis with elevation of serum estrone and androstenedione as well as urine chorionic gonadotropin. The fine structural features of the neoplasm resembled those of nontumorous Leydig cells as well
D S King et al.
JAMA, 281(21), 2020-2028 (1999-06-08)
Androstenedione, a precursor to testosterone, is marketed to increase blood testosterone concentrations as a natural alternative to anabolic steroid use. However, whether androstenedione actually increases blood testosterone levels or produces anabolic androgenic effects is not known. To determine if short-
Floriane Guibert et al.
Proceedings. Biological sciences, 280(1753), 20122368-20122368 (2012-12-21)
The prenatal environment is a source of phenotypic variability influencing the animal's characteristics. Prenatal stress affects not only the development of offspring, but also that of the following generation. Such effects have been best documented in mammals but can also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service