Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C23H39NO19
CAS Number:
Molecular Weight:
633.55
PubChem Substance ID:
UNSPSC Code:
12352201
Beilstein/REAXYS Number:
75357
MDL number:
biological source
human milk
assay
≥98% (HPAE/PAD)
storage temp.
−20°C
SMILES string
[Na+].[H]C(=O)[C@H](O)[C@@H](O)[C@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O[C@@]2(C[C@H](O)[C@@H](NC(C)=O)[C@@H](O2)[C@H](O)[C@H](O)CO)C([O-])=O)[C@H]1O)[C@H](O)CO
InChI
1S/C23H39NO19.Na/c1-7(29)24-13-8(30)2-23(22(38)39,42-19(13)15(35)10(32)4-26)43-20-16(36)12(6-28)40-21(17(20)37)41-18(11(33)5-27)14(34)9(31)3-25;/h3,8-21,26-28,30-37H,2,4-6H2,1H3,(H,24,29)(H,38,39);/q;+1/p-1/t8-,9-,10+,11+,12+,13+,14+,15+,16-,17+,18+,19+,20-,21-,23-;/m0./s1
InChI key
LTWFUJWFLMHANB-TZCPRLTCSA-M
Application
3′-Sialyllactose is a compound wherein the acetylneuraminyl (NANA) unit is connected to the galactosyl unit of lactose at the 3 position. In 6′-sialylactose, this connection is at the 6 position. 3′-Sialyllactose and 6′-Sialyllactose are used to differentiate and characterize binding domains of viruses, such as influenza and rhinitis viruses, that recognize NANA capped cell surface receptors. 3′-Sialyllactose and 6′-sialylactose are used as reference compounds in analytical methods developed to measure their presence in materials such as milk and colostrums.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.