Skip to Content
Merck
All Photos(1)

Documents

A9074

Sigma-Aldrich

2-Amino-2-methyl-1,3-propanediol

BioXtra, pH 10.0-12.0 (20 °C, 0.5 M in H2O), ≥99%

Synonym(s):

AMPD, Ammediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2C(NH2)CH3
CAS Number:
Molecular Weight:
105.14
Beilstein:
635708
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

vapor density

3.63 (vs air)

vapor pressure

~10 mmHg ( 152 °C)

product line

BioXtra

Assay

≥99%

impurities

<0.005% Phosphorus (P)
<0.1% Insoluble matter

ign. residue

<0.1%

pH

10.0-12.0 (20 °C, 0.5 M in H2O)

useful pH range

7.8-9.7

pKa (25 °C)

8.8

bp

151 °C/10 mmHg (lit.)

mp

100-110 °C (lit.)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): <0.05%
sulfate (SO42-): <0.05%

cation traces

Al: <0.001%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.005%
Pb: <0.001%
Zn: <0.0005%

SMILES string

CC(N)(CO)CO

InChI

1S/C4H11NO2/c1-4(5,2-6)3-7/h6-7H,2-3,5H2,1H3

InChI key

UXFQFBNBSPQBJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Buffer component in a SDS-gradient gel electrophoresis system that separates polypeptides in the molecular weight range of 1500 to 100,000. Used as a spacer in isotachophoresis of proteins. Also used as a buffer for the determination of alkaline phosphatase activity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karen M O'Connor et al.
Journal of pharmaceutical sciences, 91(10), 2271-2281 (2002-09-13)
Dissolution of diclofenac from compressed discs containing mixtures of a diclofenac salt and a basic excipient, in various w/w ratios, was examined. Two diclofenac salts, diclofenac deanol (DDNL) and diclofenac tert-butylamine, and the basic excipient 2-amino-2-methyl-1,3-propanediol (AMPD) were examined. Inclusion
H Kikuchi et al.
The Tohoku journal of experimental medicine, 173(4), 391-397 (1994-08-01)
The method by MacQueen and Plaut was modified to devise simple microdetermination method of l-lactate which is applicable to deproteinized blood sample. Blood was deproteinized with addition of 25% (w/w) trichloroacetic acid (TCA), and 0.075 M 2-amino-2-methyl-1, 3-propanediole (AMPD) was
Richard E Wooley et al.
Diseases of aquatic organisms, 59(3), 263-267 (2004-07-22)
The antimicrobial agents used to treat bacterial fish diseases are archaic, and their uses may result in the emergence of drug-resistant bacterial strains. This study evaluated the in vitro antimicrobial activity of combinations of Tricide and neomycin or oxytetracycline on
Jung-Yeon Park et al.
Environmental science & technology, 37(8), 1670-1675 (2003-05-07)
Acid gas absorption technology is of great importance in these days for the prevention of global warming and the resulting worldwide climate change. More efficient process design and development for the removal of acid gases has become important, together with
K Jung et al.
Clinica chimica acta; international journal of clinical chemistry, 102(2-3), 215-219 (1980-03-28)
The inhibitory effect of inorganic phosphate on the activity determination of isoenzymes of alkaline phosphatase (AP) in diethanolamine (DEA), glycine and 2-amino-2-methyl-1,3-propandiol (AMPD) buffer was studied. This effect depends on the buffer used and isoenzyme investigated. Especially the placental isoenzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service