Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C9H18N2O3
CAS Number:
Molecular Weight:
202.25
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(C)N)C(O)=O
InChI
1S/C9H18N2O3/c1-5(2)4-7(9(13)14)11-8(12)6(3)10/h5-7H,4,10H2,1-3H3,(H,11,12)(H,13,14)
InChI key
RDIKFPRVLJLMER-UHFFFAOYSA-N
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.