Skip to Content
Merck
All Photos(1)

Key Documents

A8798

Sigma-Aldrich

Arachidonic acid sodium salt

from Porcine liver, ≥99% (capillary GC), waxy solid

Synonym(s):

5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid, Sodium arachidonate, cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid sodium salt, 20:4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31O2Na
CAS Number:
Molecular Weight:
326.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

Porcine liver

Assay

≥99% (capillary GC)

form

waxy solid

color

white to off-white

solubility

methanol: 50 mg/mL, clear, colorless to faintly yellow

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC([O-])=O

InChI

1S/C20H32O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22;/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3,(H,21,22);/q;+1/p-1/b7-6-,10-9-,13-12-,16-15-;

InChI key

DDMGAAYEUNWXSI-XVSDJDOKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Arachidonic acid (AA) is a polyunsaturated ω6 fatty acid constituent of the phospholipids of cell membranes. Structurally, it is a 20-carbon chain that contains four cis double bonds, which allow the free acid form of AA to be an insoluble oil; this can be converted to the water-soluble sodium salt form within the normal physiological pH range.[1] AA is metabolized by multiple enzymes to yield various metabolites; AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation and apoptosis.

Application

Arachidonic acid is used in assays to measure activity of oxidizing enzymes, such as cyclooxygenase, as well as in assays to determine platelet inhibition in whole blood. AA has been demonstrated to bind to the α subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs), to have a role in the development and progression of prostate cancer, and to enhance formation of lipid bodies in human mast cells.[2]

Biochem/physiol Actions

Released arachidonic acid (AA) reacts with molecular oxygen nonenzymatically, through oxidative stress, and enzymatically through the action of oxygenase enzymes.[1] AA is oxidized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism. Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.

Caution

The sodium salts of arachidonic acid are very sensitive to oxidation and will turn yellow and deteriorate rapidly in air. They are packaged under vacuum in sealed ampules. Once opened the product must be used quickly or transferred into an inert atmosphere (dry argon) as soon as possible.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tony L Walker et al.
Frontiers in behavioral neuroscience, 4 (2010-08-31)
Conditioned inhibition (CI) is a major category of associative learning that occurs when an organism learns that one stimulus predicts the absence of another. In addition to being important in its own right, CI is interesting because its occurrence implies
Arachidonic acid as a bioactive molecule.
A R Brash
The Journal of clinical investigation, 107(11), 1339-1345 (2001-06-08)
Joel S Cavallo et al.
Frontiers in behavioral neuroscience, 8, 354-354 (2014-11-07)
Extinction of a conditioned association is typically viewed as the establishment of new learning rather than the erasure of the original memory. However, recent research in the nudibranch, Hermissenda crassicornis (H.c.) demonstrated that extinction training (using repeated light-alone presentations) given
Andrea Dichlberger et al.
Journal of lipid research, 52(12), 2198-2208 (2011-10-06)
Lipid droplets, also called lipid bodies (LB) in inflammatory cells, are important cytoplasmic organelles. However, little is known about the molecular characteristics and functions of LBs in human mast cells (MC). Here, we have analyzed the genesis and components of
Christo Z Christov et al.
Biophysical journal, 104(5), L5-L7 (2013-03-12)
A key step in the cyclooxygenase reaction cycle of cyclooxygenase 1 (COX-1) is abstraction of the pro-S hydrogen atom of the arachidonic acid by a radical that is formed at the protein residue Tyr-385. Here we investigate this reaction step

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service