Skip to Content
Merck

Skip To

A8171

D-Ala-Leu-Lys-7-amido-4-methylcoumarin

≥95% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 162,00

About This Item

Empirical Formula (Hill Notation):
C25H37N5O5
CAS Number:
Molecular Weight:
487.59
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:

€ 162,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(C)N)C(=O)NC(CCCCN)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C25H37N5O5/c1-14(2)11-20(30-23(32)16(4)27)25(34)29-19(7-5-6-10-26)24(33)28-17-8-9-18-15(3)12-22(31)35-21(18)13-17/h8-9,12-14,16,19-20H,5-7,10-11,26-27H2,1-4H3,(H,28,33)(H,29,34)(H,30,32)

InChI key

FHYIXLQAMONDNN-UHFFFAOYSA-N

General description

Fluorogenic substrate for plasmin.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B4636S9761S8758
assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98%

solubility

water: 1 mg/mL, clear, colorless

solubility

ethanol: 2 mg/mL, clear, colorless

solubility

methanol: 50 mg/mL, clear, colorless

solubility

50% acetic acid: 1 mg/mL, clear, colorless

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters