Skip to Content
Merck
All Photos(1)

Key Documents

A7015

Sigma-Aldrich

N-(2-Fluorenyl)acetamide

≥98% (HPLC)

Synonym(s):

2-AAF, 2-Acetamidofluorene, N-Acetyl-2-aminofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
Beilstein:
2807677
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

mp

192-196 °C (lit.)

SMILES string

CC(=O)Nc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C15H13NO/c1-10(17)16-13-6-7-15-12(9-13)8-11-4-2-3-5-14(11)15/h2-7,9H,8H2,1H3,(H,16,17)

InChI key

CZIHNRWJTSTCEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-Fluorenyl)acetamide (2-Acetamidofluorene, 2-AAF), a genotoxic carcinogen, is used to induce liver cancer in animal models such as the 2-AAF/partial hepatectomy rat. 2-AAF may be used to study the mechanism of liver carcinogenesis and as a reference material during its identification or quantitation.

Biochem/physiol Actions

A genotoxic carcinogen that is used to model liver carcinogenesis in rat. When N-hydroxylated by cytochrome CYP1A2 in the liver, 2-AAF forms adducts with DNA and is tumorigenic in liver and bladder.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Xiang et al.
PloS one, 7(4), e35180-e35180 (2012-04-20)
Macrophages are known to play an important role in hepatocyte mediated liver regeneration by secreting inflammatory mediators. However, there is little information available on the role of resident macrophages in oval cell mediated liver regeneration. In the present study we
Samrat Chakraborty et al.
Molecular therapy. Nucleic acids, 20, 34-49 (2020-03-09)
Site-specific delivery of chemotherapeutics specifically to neoplastic hepatocytes without affecting normal hepatocytes should be a focus for potential therapeutic management of hepatocellular carcinoma (HCC). The aptamer TLS 9a with phosphorothioate backbone modifications (L5) has not been explored so far for
Mohammed A El-Magd et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 114, 108732-108732 (2019-03-30)
Pretreatment of mesenchymal stem cells (MSCs) with melatonin (Mel) improves their potential therapeutic effect on chronic diseases and cancers. However, this preconditioning strategy may direct the effect of Mel toward MSCs alone and deprive cancer cells of the oncostatic effect
Sana Javaid Awan et al.
Cell biology international, 41(1), 51-61 (2016-11-03)
Hepatic oval cells are likely to be activated during advanced stage of liver fibrosis to reconstruct damaged hepatic tissue. However, their scarcity, difficulties in isolation, and in vitro expansion hampered their transplantation in fibrotic liver. This study was aimed to
Bu-Gao Zhou et al.
Frontiers in pharmacology, 9, 1165-1165 (2018-11-09)
It is known that excessive hepatocellular apoptosis is a typical characteristic of hepatic disease, and is regulated by the mammalian target of rapamycin (mTOR) signaling pathway. As the main active component of Kudzu (Pueraria lobata) roots, which is frequently used

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service