Skip to Content
Merck

Skip To

A6659

AMPSO

≥99% (titration)

Synonym(s):

N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 78,20
100 g
Check Cart for Availability
€ 216,00

About This Item

Empirical Formula (Hill Notation):
C7H17NO5S
CAS Number:
Molecular Weight:
227.28
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
269-991-7
Beilstein/REAXYS Number:
5929916
MDL number:

€ 78,20


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (titration)

form

crystalline

useful pH range

8.3-9.7

pKa (25 °C)

9.0

solubility

water: 0.333 g/mL, clear to slightly hazy, colorless

application(s)

diagnostic assay manufacturing

SMILES string

CC(C)(CO)NCC(O)CS(O)(=O)=O

InChI

1S/C7H17NO5S/c1-7(2,5-9)8-3-6(10)4-14(11,12)13/h6,8-10H,3-5H2,1-2H3,(H,11,12,13)

InChI key

ACERFIHBIWMFOR-UHFFFAOYSA-N

General description

N-(1,1-dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulfonic acid (AMPSO) is used as commercial pH buffers.

Application

AMPSO has been used in electroblotting the electrophoresed protein to nitrocellulose.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A1911M8389T9269
assay

≥99% (titration)

assay

-

assay

≥99% (titration)

assay

≥99% (titration)

Quality Level

200

Quality Level

200

Quality Level

400

Quality Level

200

form

crystalline

form

crystalline powder

form

crystalline powder

form

crystalline powder

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

solubility

water: 0.333 g/mL, clear to slightly hazy, colorless

solubility

water: 0.33 g/mL, clear to slightly hazy, colorless

solubility

water: 0.333 mg/mL, clear, colorless

solubility

warm water: 0.333 g/mL, clear to slightly hazy, colorless

useful pH range

8.3-9.7

useful pH range

8.3-9.7

useful pH range

6.2-7.6

useful pH range

7.0-8.2


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters