Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C21H25ClN2OS · HCl
CAS Number:
Molecular Weight:
425.41
UNSPSC Code:
41106305
NACRES:
NA.77
MDL number:
eCl@ss:
32110600
Assay:
≥98% (HPLC)
Form:
solid
assay
≥98% (HPLC)
form
solid
color
white
mp
175.2-176.7 °C
solubility
H2O: 23 mg/mL
originator
Abbott
storage temp.
2-8°C
General description
Guanylyl cyclases are signal transduction enzyme that convert GTP to cyclic GMP. Soluble guanylate cyclase is activated by a gaseous ligand, nitric oxide.. The nitric oxide activation of soluble guanylate, resulting in GTP conversion to GMP, is important in physiological processes such as as smooth muscle relaxation, neurotransmission, inhibition of platelet aggregation and immune response. A-350619 is an activator of soluble guanylyl cyclase.
Application
A-350619 relaxes cavernosum tissue strips in a dose dependent manner with EC50 of 80 μMM. Because A-350619 at 1 μMmol/kg induces penile erection in a conscious rat model, it may be useful as an alternate method of enhancing the effect of nitric oxide for the treatment of sexual dysfunction.
Biochem/physiol Actions
Features and Benefits
This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Legal Information
Sold under license from Abbott Laboratories
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


