Skip to Content
Merck

A6458

Acetonitrile solution

20% in H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
UNSPSC Code:
12352103
Beilstein/REAXYS Number:
4124158
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


concentration

20% in H2O

density

0.782 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Acetonitrile solution


pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library