Skip to Content
Merck
All Photos(2)

Documents

A5560

Sigma-Aldrich

Anti-Water Channel Aquaporin 1 antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot (chemiluminescent): 1:400-1:800

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AQP1(358)
rat ... Aqp1(25240)

General description

Aquaporins are transmembrane proteins composed of 6 α helices that regulate the flow of water across the membrane. AQP1 is highly expressed on the apical membrane of kidneys.
Rabbit Anti-Water Channel Aquaporin 1 antibody binds to human and rat aquaporin 1.

Immunogen

synthetic peptide corresponding to amino acids 243-261 of human aquaporin 1 conjugated to KLH with glutaraldehyde. The epitope is identical in human, rodent, and cattle and only slightly different in frog.

Application

Rabbit Anti-Water Channel Aquaporin 1 antibody has been used for immunocytochemical assays. The product can also be used for immunohistochemistry and western blot.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, and 0.025% sodium azide

Preparation Note

Isolated on immobilized Protein A

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R T Boyle et al.
Bulletin of environmental contamination and toxicology, 90(3), 387-390 (2013-01-01)
We used immunocytochemical and fluorometric techniques to show that gill cells of two marine invertebrates, the crab Neohelice granulata (osmoregulator) and the clam Mesodesma mactroides (osmoconformer), increase the expression of membrane transporters [Na(+)/K(+)-ATPase and aquaporin (AQP1)] after whole-animals exposure (96 h)
Kamila Foguesatto et al.
Cell biology international, 46(6), 976-985 (2022-03-09)
The physiological variations during the crustacean molting cycle have intrigued researchers for many years. Maintaining osmotic homeostasis in the face of hemolymph dilution and dealing with dynamic intracellular and extracellular calcium fluctuations are challenges these animals continuously confront. It has
Elisabeth Kruse et al.
Genome biology, 7(2), 206-206 (2006-03-09)
Water is the major component of all living cells, and efficient regulation of water homeostasis is essential for many biological processes. The mechanism by which water passes through biological membranes was a matter of debate until the discovery of the
Esperanza Gonzalez et al.
International journal of biological sciences, 17(8), 1864-1877 (2021-06-17)
Alcohol abuse has a high impact on the mortality and morbidity related to a great number of diseases and is responsible for the development of alcoholic liver disease (ALD). It remains challenging to detect and evaluate its severity, which is
Sidra Khan et al.
Cancers, 15(18) (2023-09-28)
Aquaporin (AQP) channels in endometrial cancer (EC) cells are of interest as pharmacological targets to reduce tumor progression. A panel of compounds, including AQP1 ion channel inhibitors (AqB011 and 5-(phenoxymethyl) furan-2-carbaldehyde, PMFC), were used to test the hypothesis that inhibition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service