Skip to Content
Merck
All Photos(1)

Key Documents

A5539

Sigma-Aldrich

AAT Media Supplement (50×) Hybri-Max

γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€63.40

€63.40


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
1 VIAL
€63.40

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

€63.40


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

sterility

γ-irradiated

product line

BioXtra

form

lyophilized powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

Reconstitution

Reconstitute contents of vial with 10 mL sterile cell culture medium. Stock solution is sufficient to prepare 500 mL medium. Final working concentration: 75 μM adenine, 0.8 μM aminopterin, 16 μM thymidine.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service