Skip to Content
Merck

Skip To

A5080

Amyloid β-Protein Fragment 22-35

≥95% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 408,00
€ 346,80

About This Item

Empirical Formula (Hill Notation):
C59H102N16O21S
CAS Number:
Molecular Weight:
1403.60
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Form:
powder
Assay:
≥95% (HPLC)

€ 346,80

List Price€ 408,00Save 15%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(NC(=O)C(C)NC(=O)CNC(=O)C(CCCCN)NC(=O)C(CC(N)=O)NC(=O)C(CO)NC(=O)CNC(=O)C(NC(=O)C(CC(O)=O)NC(=O)C(N)CCC(O)=O)C(C)C)C(C)CC)C(=O)NCC(=O)NC(CC(C)C)C(=O)NC(CCSC)C(O)=O

InChI

1S/C59H102N16O21S/c1-11-30(7)47(57(93)65-25-42(79)67-36(21-28(3)4)52(88)70-35(59(95)96)18-20-97-10)75-58(94)48(31(8)12-2)74-49(85)32(9)66-41(78)24-63-51(87)34(15-13-14-19-60)69-53(89)37(22-40(62)77)72-55(91)39(27-76)68-43(80)26-64-56(92)46(29(5)6)73-54(90)38(23-45(83)84)71-50(86)33(61)16-17-44(81)82/h28-39,46-48,76H,11-27,60-61H2,1-10H3,(H2,62,77)(H,63,87)(H,64,92)(H,65,93)(H,66,78)(H,67,79)(H,68,80)(H,69,89)(H,70,88)(H,71,86)(H,72,91)(H,73,90)(H,74,85)(H,75,94)(H,81,82)(H,83,84)(H,95,96)

InChI key

KUDAPIFTNDCCFG-UHFFFAOYSA-N

Gene Information

human ... APP(351)

Biochem/physiol Actions

Has cytotoxic effect on cultured neurons from rat hippocampus in serum free medium. Also forms aggregates and fibrils similar to those of β-amyloid protein in senile plaque cores.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2326A4723A4848
assay

≥95% (HPLC)

assay

≥90% (HPLC)

assay

≥90% (HPLC)

assay

≥87% (HPLC)

form

powder

form

powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Gene Information

human ... APP(351)

Gene Information

human ... APP(351)

Gene Information

human ... APP(351)

Gene Information

human ... APP(351)

UniProt accession no.

P05067

UniProt accession no.

P05067

UniProt accession no.

P05067

UniProt accession no.

P05067

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters