Skip to Content
Merck

A5004

2-Amino-4,6-dimethylpyrimidine

Synonym(s):

Acetylacetoneguanidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H9N3
CAS Number:
Molecular Weight:
123.16
EC Number:
212-181-5
UNSPSC Code:
12352204
MDL number:
Beilstein/REAXYS Number:
110735
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mp

151-153 °C (lit.)

SMILES string

Cc1cc(C)nc(N)n1

InChI

1S/C6H9N3/c1-4-3-5(2)9-6(7)8-4/h3H,1-2H3,(H2,7,8,9)

InChI key

IDQNBVFPZMCDDN-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library