Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352202
Assay:
~95%
Form:
solid
assay
~95%
form
solid
storage temp.
2-8°C
Other Notes
A mixture of structural isomers TB1 and TB2.1
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 2 Oral - Repr. 1B
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

