Skip to Content
Merck

Skip To

A4564

LS-Tetra saccharide a

from human milk

Synonym(s):

α-NeuNAc-(2→3)-β-Gal-(1→3)-b-GlcNAc-(1→3]-β-Gal-(1→4)-Glc, LST a, N-Acetylneuramin-lacto-N-tetraose a

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C37H62N2O29
CAS Number:
Molecular Weight:
998.88
UNSPSC Code:
12352200
MDL number:


biological source

human milk

storage temp.

−20°C

SMILES string

CC(=O)NC1C(O)CC(OC2C(O)C(CO)OC(OC3C(O)C(CO)OC(OC4C(O)C(CO)OC(OC(C(O)CO)C(O)C(O)C=O)C4O)C3NC(C)=O)C2O)(OC1C(O)C(O)CO)C(O)=O





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library