Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C22H33N3O5S
CAS Number:
Molecular Weight:
451.58
UNSPSC Code:
12352200
MDL number:
assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CSCCC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(Cc1ccccc1)C(O)=O
Application
Used in studies on the structural requirements and specificity of synthetic peptide chemoattractants.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.