Skip to Content
Merck

Key Documents

Skip To

A3913

L-Argininamide dihydrochloride

≥98%, suitable for ligand binding assays

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H15N5O · 2HCl
CAS Number:
Molecular Weight:
246.14
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Argininamide dihydrochloride,

assay

≥98%

Quality Segment

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.N[C@@H](CCCNC(N)=N)C(N)=O

InChI

1S/C6H15N5O.ClH/c7-4(5(8)12)2-1-3-11-6(9)10;/h4H,1-3,7H2,(H2,8,12)(H4,9,10,11);1H/t4-;/m0./s1

InChI key

BPQLYFCEVVKLLX-WCCKRBBISA-N

Application

L-Argininamide dihydrochloride has been used as an additive to compare its effect over L-arg in egg lysozyme refolding studies. It has also been used as a component of substrate stock solution for enzymatic kyotorphin synthesis.

Biochem/physiol Actions

L-Argininamide (L-Arm) is a hydrophilic amino acid derivative that is used as a model compound in physicochemical characteristic studies of ligand binding DNA aptamers and their potential development as fluorescent aptasensors.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T4626A3785C3647
description

-

description

-

description

≥98%

description

-

form

powder

form

powder

form

powder

form

powder

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

technique(s)

ligand binding assay: suitable

assay

≥98%

assay

≥98% (TLC)

assay

≥98%

assay

≥98% (TLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200

color

white to off-white

color

white

color

-

color

white


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 1

Target Organs

Eyes

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters