Skip to Content

A3261

Arg-Arg β-naphthylamide trihydrochloride

≥99% (HPLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H33N9O2 · 3HCl
CAS Number:
Molecular Weight:
564.94
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Arg-Arg β-naphthylamide trihydrochloride,

assay

≥99% (HPLC)

Quality Segment

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

Cl.Cl.Cl.N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)Nc1ccc2ccccc2c1

InChI

1S/C22H33N9O2.3ClH/c23-17(7-3-11-28-21(24)25)19(32)31-18(8-4-12-29-22(26)27)20(33)30-16-10-9-14-5-1-2-6-15(14)13-16;;;/h1-2,5-6,9-10,13,17-18H,3-4,7-8,11-12,23H2,(H,30,33)(H,31,32)(H4,24,25,28)(H4,26,27,29);3*1H/t17-,18-;;;/m0.../s1

InChI key

RNVXXFQJTJQVER-NCXIRTITSA-N

General description

Arg-Arg β-naphthylamide is a dipeptide and a β-naphthylamide derivative.
Substrate for dipeptidyl aminopeptidase III.

Application

Arg-Arg β-naphthylamide trihydrochloride has been used as a substrate for the enzyme dipeptidyl peptidase 3 expressed in human embryonic kidney cells (HEK293T) and as a substrate for peptidase from black-pigmented bacteroides (BPB) and spirochetes.


pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library