Skip to Content
Merck
All Photos(1)

Documents

A3145

Sigma-Aldrich

Apigenin

≥97% (TLC), from parsley, powder

Synonym(s):

4′,5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
262620
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

parsley

Assay

≥97% (TLC)

form

powder

color

yellow

mp

>300 °C (lit.)

solubility

DMSO: 27 mg/mL
1 M KOH: 50 mg/mL

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=CC(=O)c3c(O)cc(O)cc3O2

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)13-7-12(19)15-11(18)5-10(17)6-14(15)20-13/h1-7,16-18H

InChI key

KZNIFHPLKGYRTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A plant flavonoid that has been found to inhibit cell proliferation by arresting the cell cycle at the G2/M phase. Inhibition of growth through cell cycle arrest and induction of apoptosis appear to be related to induction of p53. Inhibits PMA-mediated tumor promotion by inhibiting protein kinase C and the resulting suppression of oncogene expression. It has also been reported to inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Émilie C Lefort et al.
Molecular nutrition & food research, 57(1), 126-144 (2012-12-01)
Apigenin (4',5,7-trihydroxyflavone, 5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one) is a flavonoid found in many fruits, vegetables, and herbs, the most abundant sources being the leafy herb parsley and dried flowers of chamomile. Present in dietary sources as a glycoside, it is cleaved in the gastrointestinal
Naiara Silva Dourado et al.
Frontiers in aging neuroscience, 12, 119-119 (2020-06-06)
Neurodegenerative disorders (ND) are characterized by the progressive and irreversible loss of neurons. Alzheimer's Disease (AD) is the most incident age-related ND, in which the presence of a chronic inflammatory compound seems to be related to its pathogenesis. Different stimuli
Benjamin Dilberger et al.
Nutrients, 11(8) (2019-08-16)
(1) Background: Polyphenols (PP) play an important role in the prevention of non-communicable diseases and may contribute to healthy aging. To investigate the molecular and cellular aspects of PP metabolites on longevity with a focus on mitochondrial function, we applied
John Sanseverino et al.
Toxicological sciences : an official journal of the Society of Toxicology, 107(1), 122-134 (2008-11-11)
Saccharomyces cerevisiae bioluminescent bioreporter assays were developed previously to assess a chemical's estrogenic or androgenic disrupting potential. S. cerevisiae BLYES, S. cerevisiae BLYAS, S. cerevisiae BLYR, were used to assess their reproducibility and utility in screening 68, 69, and 71
Kaihui Lu et al.
Journal of natural products, 76(4), 672-678 (2013-03-26)
Adipocyte dysfunction is a major cause of obesity, which is associated strongly with many disorders including psychological and medical morbidities, metabolic abnormalities, and cardiovascular diseases as well as a series of cancers. This study investigated the antiadipogenic activity of scutellarin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service