Skip to Content
Merck
All Photos(1)

Key Documents

A2643

Sigma-Aldrich

α-Amylase from porcine pancreas

Type VII-A, DFP treated, ammonium sulfate suspension, ≥500 units/mg protein (E1%/280)

Synonym(s):

β-N-acetylglucosaminidase porcine placenta, PPA, al1,4 glucan-4-glucanohydrolase,, porcine pancreas α-amylase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

Pricing and availability is not currently available.

type

Type VII-A

form

ammonium sulfate suspension

specific activity

≥500 units/mg protein (E1%/280)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Molecular mass: 51-54 kDa.
α-Amylase isolated from porcine pancreas is a glycoprotein. It is a single polypeptide chain of ~475 residues containing two SH groups and four disulfide bridges and a tightly bound Ca2+ necessary for stability. Chloride ions are necessary for activity and stability. The pH range for activity is 5.5 to 8.0, with the pH optimum at 7.

Unit Definition

One unit will liberate 1.0 mg of maltose from starch in 3 min at pH 6.9 at 20 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4, pH 6.1

inhibitor

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fushan Liu et al.
Journal of experimental botany, 60(15), 4423-4440 (2009-10-07)
The amylose extender (ae(-)) mutant of maize lacks starch branching enzyme IIb (SBEIIb) activity, resulting in amylopectin with reduced branch point frequency, and longer glucan chains. Recent studies indicate isozymes of soluble starch synthases form high molecular weight complexes with
Buford L Nichols et al.
The Journal of nutrition, 139(4), 684-690 (2009-02-06)
Starch is the major source of food glucose and its digestion requires small intestinal alpha-glucosidic activities provided by the 2 soluble amylases and 4 enzymes bound to the mucosal surface of enterocytes. Two of these mucosal activities are associated with
Wiesław Jerzy Cubała et al.
Journal of affective disorders, 157, 14-17 (2014-03-04)
Altered monoamine neurotransmission accompanied by hypothalamic-pituitary-adrenal axis dysfunction and autonomic nervous system hyperactivity have been associated with major depressive disorder (MDD). Salivary α-amylase (sAA) is indicative of autonomic activation and reflects central noradrenergic activity. Scarce studies on sAA in MDD
Heidemarie K Laurent et al.
Psychosomatic medicine, 75(8), 744-750 (2013-10-01)
Neurotrophins such as nerve growth factor (NGF) may represent a stress-responsive system complementing the better known neuroendocrine (hypothalamic-pituitary-adrenal axis) and autonomic nervous system, but there is little evidence for NGF response to acute stress in humans because noninvasive measures have
Phuong Lan Tran et al.
FEBS letters, 588(4), 620-624 (2014-01-21)
The action pattern of Bacillus licheniformis thermostable α-amylase (BLA) was analyzed using a series of (14)C-labeled and non-labeled maltooligosaccharides from maltose (G2) to maltododecaose (G12). Maltononaose (G9) was the preferred substrate, and yielded the smallest Km=0.36 mM, the highest kcat=12.86

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service