A2295
Acivicin
Synonym(s):
α-Amino-3-chloro-4,5-dihydro-5-isoxazoleacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H7ClN2O3
CAS Number:
Molecular Weight:
178.57
MDL number:
UNSPSC Code:
12352200
Recommended Products
storage temp.
2-8°C
SMILES string
N[C@@H]([C@@H]1CC(Cl)=NO1)C(O)=O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service