Skip to Content
Merck

Skip To

A2145

Acridine hydrochloride

Synonym(s):

Acridine monohydrochloride, Acridinium chloride, acridin-10-ium chloride

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C13H9N · HCl
CAS Number:
Molecular Weight:
215.68
EC Number:
241-762-6
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
3697519
MDL number:


mp

250-255 °C (dec.)

solubility

H2O: soluble, methanol: soluble

SMILES string

Cl[H].c1ccc2nc3ccccc3cc2c1

InChI

1S/C13H9N.ClH/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)14-12;/h1-9H;1H

InChI key

XUESTGHCVFYOLL-UHFFFAOYSA-N

Application

Acridine hydrochloride is a fluorescent compound with λexcitation and λemission of 355 nm and 440 nm, respectively. To investigate the release properties of multilayer nanocapsules, acridine hydrochloride has been utilized as a hydrophilic drug model and was loaded into the nano-structures .


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves

target_organs

Respiratory system



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library