Skip to Content
Merck
All Photos(1)

Key Documents

A1980

Sigma-Aldrich

A3 hydrochloride

≥98% (HPLC), solid

Synonym(s):

N-(2-Aminoethyl)-5-chloronaphthalene-1-sulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€142.00

€142.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€142.00

About This Item

Empirical Formula (Hill Notation):
C12H13ClN2O2S · HCl
CAS Number:
Molecular Weight:
321.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€142.00


Estimated to ship on22 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: 24 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].NCCNS(=O)(=O)c1cccc2c(Cl)cccc12

InChI

1S/C12H13ClN2O2S.ClH/c13-11-5-1-4-10-9(11)3-2-6-12(10)18(16,17)15-8-7-14;/h1-6,15H,7-8,14H2;1H

InChI key

VWAGIWCLJAQLAL-UHFFFAOYSA-N

Biochem/physiol Actions

Non-selective casein kinase (CK) inhibitor. A shorter alkyl chain derivative of W-7 that inhibits casein kinase I (Ki = 80 μM), casein kinase II (Ki = 5.1 μM), myosin light chain kinase (Ki = 7.4 μM), protein kinase A (Ki = 4.3 μM), protein kinase C (Ki= 47 μM), and protein kinase G (Ki = 3.8 μM).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service