Skip to Content
Merck

A170

Aminorex

solid

Synonym(s):

4,5-Dihydro-5-phenyl-2-oxazolamine, Aminoxaphen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H10N2O
CAS Number:
Molecular Weight:
162.19
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

drug control

USDEA Schedule I; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada, kontrollierte Droge in Deutschland

color

white

solubility

0.1 M HCl: soluble, H2O: insoluble, ethanol: soluble

SMILES string

NC1=NCC(O1)c2ccccc2

InChI

1S/C9H10N2O/c10-9-11-6-8(12-9)7-4-2-1-3-5-7/h1-5,8H,6H2,(H2,10,11)

InChI key

SYAKTDIEAPMBAL-UHFFFAOYSA-N

General description

Anorexic; a substrate of the serotonin transporter that also releases serotonin from platelets.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


S Friström et al.
Acta pharmacologica et toxicologica, 41(3), 218-224 (1977-09-01)
The effect on 5-hydroxytryptamine release from rabbit platelets to plasma of ten known sympathomimetic or anorectic phenethylamines and eight N- or O-acetyl derivatives of these substances were studied in order to find possible structure-action relationships and a possible correlation to
E K Weir et al.
Circulation, 94(9), 2216-2220 (1996-11-01)
The appetite suppressant aminorex fumarate is thought to have caused an epidemic of pulmonary hypertension in Europe in the 1960s. More recently, pulmonary hypertension has been described in some patients taking other amphetamine-like, anorexic agents: fenfluramine and its d-isomer, dexfenfluramine.
S M Paul et al.
Science (New York, N.Y.), 218(4571), 487-490 (1982-10-29)
Saturable and stereospecific binding sites for (+)-[3H]amphetamine were demonstrated in membrane preparations from rat brain. The density of these binding sites varies among brain regions and is highest in the hypothalamus and brainstem. Specific (+)-[3H]amphetamine binding in hypothalamus is largely