Skip to Content
Merck
All Photos(1)

Key Documents

A167

Sigma-Aldrich

D(−)-2-Amino-7-phosphonoheptanoic acid

solid

Synonym(s):

D-AP-7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16NO5P
CAS Number:
Molecular Weight:
225.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

H2O: soluble

SMILES string

N[C@H](CCCCCP(O)(O)=O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective NMDA glutamate receptor antagonist; anticonvulsant; active enantiomer of 2-amino-7-phosphonoheptanoic acid.

Caution

Hygroscopic. Store desiccated at RT.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Prigol et al.
Chemico-biological interactions, 193(1), 65-70 (2011-05-31)
The present study investigated the role of the glutathione system in seizures induced by diphenyl diselenide (PhSe)(2) (50 mg/kg) in rat pups (post natal day, 12-14). Reduced glutathione (GSH) (300 nmol/site; i.c.v.), administered 20 min before (PhSe)(2), abolished the appearance
Daniele Cristina Aguiar et al.
Journal of neuroscience research, 87(11), 2418-2429 (2009-04-11)
Innate fear stimulus induces activation of neurons containing the neuronal nitric oxide synthase enzyme (nNOS) in defensive-related brain regions such as the dorsolateral periaqueductal gray (dlPAG). Intra-dlPAG administration of nitric oxide synthase (NOS) inhibitors and glutamate antagonists induce anxiolytic-like responses.
M Benveniste et al.
British journal of pharmacology, 104(1), 207-221 (1991-09-01)
1. The kinetics of action of 17 structurally related NMDA receptor competitive antagonists were measured under voltage clamp in mouse hippocampal neurones. Analysis of the response to rapid changes in antagonist concentration during constant application of agonist was used to
Luiz F S Souza-Pinto et al.
Pharmacology, biochemistry, and behavior, 87(2), 250-257 (2007-06-01)
It is well established that the most persistent sign of withdrawal from chronic benzodiazepine use in humans is anxiety. In contrast to other types of drugs of abuse, the emergence of this anxiety does not seem to be linked directly
Abbas Haghparast et al.
European journal of pain (London, England), 11(8), 855-862 (2007-02-13)
The nucleus cuneiformis (CnF), located just ventrolateral to the periaqueductal gray, is part of the descending pain modulatory system. Neurons in the CnF project to medullary nucleus raphe magnus (NRM), which plays an important role on pain modulation. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service